Learn More
Sartorius Arium™ Tubing Extension Set
Product Code. 17214233 Shop All Sartorius Products
Click to view available options
For Use With (Equipment):
Arium Smart Station Ultrapure Water
Arium™ Smart Station Pure Water
Unit Size:
1 unit
Product Code. 17214233
Brand: Sartorius H2OATESUP
Only null left
Add to basket
Add to basket
Extension set (4 m) for Arium™ Smart Station, used for uncomplicated extension of connection between Arium™ Smart Station and connected water system
  • Extended tubing set used for Smart Station ultrapure and pure for uncomplicated extension of standard supplied tubing set
  • Operating range can be easily increased from 2 to 4 m
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Arium Smart Station Ultrapure Water
Type Tubing