missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ ARN 14494

Product Code. 18765736
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18765736 50 mg 50mg
18718176 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18765736 Supplier R&D Systems™ Supplier No. 7134/50
  • ONLINE EXCLUSIVE PRICE
    791.89 €
    714.00 € / 50mg
    Save 77.89 €
    9% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent serine palmitoyltransferase inhibitor

ARN 14494 is a potent serine palmitoyltransferase inhibitor (IC50 = 27.3 nM). ARN 14494 inhibits synthesis of long chain ceramides and dihydroceramides in an in vitro model of Alzheimer's diease. The compound prevents the synthesis of proinflammatory cytokines and the oxidative stress-related enzymes iNOS and COX-2 in mouse primary cortical astrocytes. ARN 14494 is neuroprotective against β-amyloid 1-42 induced neurotoxicity in primary cortical neurons co-cultured with astrocytes.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rel-1-[1-[2-[(3 aR,6 AS)-Hexahydrocyclopenta[c]pyrrol-2(1 H)-yl]-2-oxoethyl]-4-piperidinyl]-2,3-dihydro-N-methyl-2-oxo-1 H-indole-5-carboxamide
CAS 1037834-43-7
Quantity 50 mg
Product Line Tocris
Target Other Transferase Inhibitors
Molecular Formula C24H32N4O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.