Learn More
Applied Biosystems™ Array Card Dye Calibration Kit
Product Code. 15800393 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
7 arrays
Unit Size:
Each
Product Code. Quantity Unit Size
15800393 7 arrays Each
1 options
Product Code. 15800393 Supplier Applied Biosystems™ Supplier No. 4432438
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Array Card Dye Calibration Kit is used to maintain your real-time PCR instrument with array card block
  • The Array Card Dye Calibration Kit is used to maintain your real-time PCR instrument with array card block. Dye calibration should be run after lamp change.
  • The kit contains FAM™, VIC™, ROX™, ROI, and background calibration plates, as well as FAM™/ROX™ and VIC™/ROX™ normalization plates.
  • Please see your instrument user guide for full instructions on use.
TRUSTED_SUSTAINABILITY
Specifications
Calibrated Dye Proprietary Mix
For Use With (Equipment) ViiA™ 7 Dx System, QuantStudio™ Dx
Product Type Array Card Dye Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Quantity 7 arrays
Calibration Type Spectral Calibration
PCR Method qPCR