Learn More
Abeba™ Arrow 8300 Shoes
Product Code. 15805081
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15805081
Brand: Abeba™ 8300/42
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Antistatic with heel patch microfiber. Sanitized™ treated.
  • CE EN ISO 20347:2012 OB FO SRA
  • Microfiber
  • White
  • Lightweight Mono-PU-outsole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Microfiber heel patch
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB FO SRA
Color White
Material Microfiber
Size 42
Style 8300