missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) iodide, 98%

Product Code. 15462127
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
15462127 10 g 10g
15472127 50 g 50g
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15462127

Brand: Thermo Scientific Alfa Aesar 045067.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 7784-45-4 | AsI3 | 455.635 g/mol

Arsenic(III) iodide is mainly used to treat rheumatism, arthritis, malaria, trypanosome infections, tuberculosis, and diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-45-4
Molecular Formula AsI3
Molecular Weight (g/mol) 455.635
MDL Number MFCD00014170
InChI Key IKIBSPLDJGAHPX-UHFFFAOYSA-N
Synonym arsenous triiodide, arsenic iii iodide, arsenic iodide, arsenic triiodide, triiodoarsine, arsenous iodide, hsdb 420, arsen iii-jodid
PubChem CID 24575
IUPAC Name triiodoarsane
SMILES [As](I)(I)I

Specifications

Boiling Point 424°C
Melting Point 140°C
Physical Form Pieces
Quantity 10 g
UN Number UN1557
Merck Index 14,803
Solubility Information Soluble in alcohol,ether,Carbon disulfide,water(6g/100 m).
Formula Weight 455.64
Percent Purity 98%
Odor Pungent
Sensitivity Moisture sensitive
Density 4.39 g/mL
Chemical Name or Material Arsenic (III) iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.