missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(Iii) Oxide, Honeywell™

Product Code. 15633270 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
125 g
Unit Size:
125g
Product Code. Quantity unitSize
15633270 125 g 125g
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15633270

Brand: Honeywell-Fluka 311383125G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

CAS 1327-53-3
Quantity 125 g
Molecular Formula As2O3
MDL Number MFCD00003433
UN Number 1561
Solubility Information Insoluble in dilute HCl ≤0.01%
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
SMILES O=[As]O[As]=O
IUPAC Name diarsorosooxidane
Molecular Weight (g/mol) 197.84
Formula Weight 197.84g/mol
Grade ACS Reagent
Residue after Ignition ≤0.02%
Chemical Name or Material Arsenic(Iii) Oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.