missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) oxide, primary standard, ACS, 99.95-100.05%

Product Code. 11357598
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11357598 100 g 100g
11367598 25 g 25g
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11357598

Brand: Thermo Scientific Alfa Aesar 033289.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 1327-53-3 | As2O3 | 197.84 g/mol

Used in the manufacturing of such products as glass and enamels. Arsenic trioxide is utilized in various studies on cellular processes, notably in investigations of leukemia and principal material for the production of many arsenic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1327-53-3
Molecular Formula As2O3
Molecular Weight (g/mol) 197.84
MDL Number MFCD00003433
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
Synonym Arsenic trioxide
IUPAC Name diarsorosooxidane
SMILES O=[As]O[As]=O

Specifications

Boiling Point 465°C (sublimation)
Melting Point 312.3°C
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.95-100.05%
UN Number UN1561
Merck Index 14,804
Solubility Information Soluble in dilute acids and alkalies. Insoluble in organic solvents. Slightly soluble in water.
Formula Weight 197.84
Percent Purity 99.95 to 100.05%
Grade ACS
Odor Odorless
Density 3.74 g/mL
Chemical Name or Material Arsenic(III) oxide, Primary standard
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.