missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) Oxide, ReagentPlus™ Grade, ≥99%, Honeywell™

Product Code. 15697620 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Glass Bottle
Unit Size:
100g
1kg
250g
25g
Product Code. Quantity Packaging unitSize
15697620 250 g Glass Bottle 250g
15607630 25 g Glass Bottle 25g
15677620 100 g Glass Bottle 100g
15687620 1 kg Glass Bottle 1kg
4 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15697620

Brand: Honeywell Research Chemicals A1010250G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 1327-53-3
Molecular Formula As2O3
Molecular Weight (g/mol) 197.84
MDL Number MFCD00003433
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
IUPAC Name diarsorosooxidane
SMILES O=[As]O[As]=O

Specifications

Boiling Point 460°C
Melting Point 313°C
Color White
Quantity 250 g
UN Number UN1561
Formula Weight 197.84g/mol
Percent Purity ≥99%
Odor Odorless
Packaging Glass Bottle
Appearance Powder
Density 3.87g/cm3 at 22.3°C
Chemical Name or Material Arsenic(III) oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.