missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) telluride, 99.999% (metals basis)

Product Code. 11388848
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388848 10 g 10g
11378848 50 g 50g
15419189 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11388848 Supplier Thermo Scientific Alfa Aesar Supplier No. 041709.09
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 12044-54-1 | As2Te3 | 532.643 g/mol

Vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12044-54-1
Molecular Formula As2Te3
Molecular Weight (g/mol) 532.643
MDL Number MFCD00014167
InChI Key GTIUFDICMGTSPM-UHFFFAOYSA-N
Synonym arsenic iii telluride, tellanylidenearsanyl tellanyl arsanetellone, arsenic tritelluride
PubChem CID 15770922
SMILES [As](=[Te])[Te][As]=[Te]

Specifications

Melting Point 381°C
Physical Form Pieces
Quantity 10 g
UN Number UN1557
Formula Weight 532.64
Percent Purity 99.999%
Packaging Plastic bottle
Specific Gravity 6.5
Density 6.5
Chemical Name or Material Arsenic(III) telluride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.