missing translation for 'onlineSavingsMsg'
Learn More

Arsenic Oxide (As2O3), TRC

Product Code. 30341291
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
30341291 10 g 10g
30331437 50 g 50g
2 options
This item is not returnable. View return policy

Product Code. 30341291

Brand: Toronto Research Chemicals TRCA77413010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Arsenic trioxide
CAS 1327-53-3
Molecular Formula As2 O3
Quantity 10 g
Synonym Arsenious trioxide, Arsenous trioxide
InChI Formula InChI=1S/As2O3/c3-1-5-2-4
SMILES O=[As]O[As]=O
IUPAC Name arsenic(3+);oxygen(2-)
Molecular Weight (g/mol) 197.84
Formula Weight 197.8279
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.