missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ART™ Barrier Specialty Pipette Tips

Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.

Brand:  Thermo Scientific™ 2330-05

Code : 03

Additional Details : Weight : 1.10250kg

Product Code. 12348452

  • 131.00 € / Pack of 960
Estimated Shipment: 02-09-2024
to see stock.
Add to basket

Description

Description

Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to aid in pipetteting for a variety of applications:

Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel

Gel loading for loading acrylamide or agarose gels

Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials

Solvent Safe for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettetes

Ergonomic pipette tips are designed for Gilson pipetteman™ pipettetes and Thermo Scientific™ Finnpipettete™ Focus pipettetes

Specifications

Specifications

200 μL
ART 200 ERGO-G
5.08cm
Rack
Gilson™ Pipetman™, Thermo Scientific™ Finnpipette™ Focus
Sterile & PCR Ready
Sterile
2 in.
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Ergonomic
ART
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers