missing translation for 'onlineSavingsMsg'
Learn More

Artemisinin, 98%

Product Code. 15466889
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466889 5 g 5g
15456889 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466889 Supplier Thermo Scientific Alfa Aesar Supplier No. J65406.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 63968-64-9 | C15H22O5 | 282.336 g/mol

Artemisinin inhibits angiogenesis by down-regulating HIF-1α and VEGF expression in mouse embryonic stem cells. Artemisinin crosses the blood-brain barrier and is an inhibitor of human NOS2 (iNOS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble to 100mM in DMSO and to 75mM in ethanol

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63968-64-9
Molecular Formula C15H22O5
Molecular Weight (g/mol) 282.336
MDL Number MFCD00081057
InChI Key BLUAFEHZUWYNDE-DKGJTOOQSA-N
Synonym artemisinine, artemisinin, gnf-pf-5341, artemisinin, 18, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.0^ 4,13 .0^ 8,13 hexadecan-10-one, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.04,13.08,13 hexadecan-10-one
PubChem CID 9838675
SMILES CC1CCC2C(C(=O)OC3C24C1CCC(O3)(OO4)C)C

Specifications

Melting Point 156°C to 157°C
Color White
Quantity 5 g
Merck Index 14,817
Solubility Information Soluble to 100mM in DMSO and to 75mM in ethanol
Optical Rotation +76° (c=0.5 in Methanol)
Formula Weight 282.33
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material Artemisinin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.