missing translation for 'onlineSavingsMsg'
Learn More

ARV-771, MedChemExpress

Product Code. 16453228
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453228

Brand: Medchem Express HY100972

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ARV-771 is a potent BET PROTAC based on E3 ligase von Hippel-Lindau with Kds of 34 nM, 4.7 nM, 8.3 nM, 7.6 nM, 9.6 nM, and 7.6 nM for BRD2(1), BRD2(2), BRD3(1), BRD3(2), BRD4(1), and BRD4(2), respectively.

ARV-771 is a potent BET PROTAC based on E3 ligase von Hippel-Lindau with Kds of 34 nM, 4.7 nM, 8.3 nM, 7.6 nM, 9.6 nM, and 7.6 nM for BRD2(1), BRD2(2), BRD3(1), BRD3(2), BRD4(1), and BRD4(2), respectively.

Specifications

Chemical Name or Material ARV-771
CAS 1949837-12-0
Color White
Purity Grade Notes Research
Molecular Formula C49H60ClN9O7S2
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (50.68 mM)
SMILES CC(C(C(C1=CC=C(Cl)C=C1)=N[C@@H](CC(NCCOCCCOCC(N[C@@H](C(C)(C)C)C(N(C[C@H](O)C2)[C@@H]2C(N[C@H](C3=CC=C(C(SC=N4)=C4C)C=C3)C)=O)=O)=O)=O)C5=NN=C(C)N65)=C6S7)=C7C
Molecular Weight (g/mol) 986.64
Formula Weight 986.64
Percent Purity 96.75%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.