missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ARV 771
GREENER_CHOICE

Product Code. 18726196
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 18726196

Brand: Tocris Bioscience™ 7256/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent BET bromodomain PROTAC™; also degrades BRD-tagged chimeric antigen receptors (CAR) in T cells

ARV 771 is a potent BET bromodomain PROTAC™ degrader (DC50 = < 1nM). Comprises a BRD4-binding moiety joined by a linker to a ligand for Von Hippel-Lindau (VHL) protein. Degrades BRD2/3/4 in castration-resistant prostate cancer (CRPC) cell lines. Reduces androgen receptor levels and induces apoptosis in CRPC cells in vitro. Down-regulates BRD4 and induces tumor regression in CRPC xenografts in mice. Also reduces leukemia burden in a mouse model. Induces degradation of BRD-tagged CAR (chimeric antigen receptor) in T cells. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2 S,4 R)-1-(-2-(tert-Butyl)-15-(-4-(4-chlorophenyl)-2,3,9-trimethyl-6 H-thieno[3,2-f][1,2,4 ]triazolo[4,3-a][1,4 ]diazepin-6-yl)-4,14-dioxo-6,10-dioxa-3,13-diazapentadecanoyl)-4-hydroxy-N-(-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carbox
CAS 1949837-12-0
Quantity 5 mg
Target Active Degraders
Molecular Formula C49H60ClN9O7S2
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.