missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AS 1517499

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AS 1517499 is a potent STAT6 inhibitor (IC50 = 21 nM). In vitro, it inhibits IL-4-induced Th2 differentiation of mouse spleen T cells (IC50 = 2.3 nM), without influencing Th1 differentiation induced by IL-12. In a mouse model of allergic bronchial asthma, treatment with AS 1517499 reduces mouse airway IL-3 production and partly inhibits the RhoA upregulation in bronchial smooth muscles.
Specifications
Specifications
| Chemical Name or Material | 2-[[2-(3-Chloro-4-hydroxyphenyl)ethyl]amino]-4-[(phenylmethyl)amino]-5-pyrimidinecarboxamide |
| CAS | 919486-40-1 |
| Quantity | 10 mg |
| Target | STAT6 |
| Molecular Formula | C20H20ClN5O2 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction