missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ AS 19

Product Code. 30001349
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001349 50 mg 50mg
30001312 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001349 Supplier R&D Systems™ Supplier No. 1968/50
  • ONLINE EXCLUSIVE PRICE
    1299.34 €
    1156.05 € / 50mg
    Save 143.29 €
    11% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent 5-HT7 agonist

AS 19 is a potent 5-HT7 receptor agonist (IC50 = 0.83 nM). Enhances memory consolidation and reverses scopolamine- or dizocilpine-induced amnesia in vivo. AS 19 also promotes melanin synthesis in B16F10 murine melanoma cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1000578-26-6
Molecular Formula C18H25N3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 283.41
Purity >98%
Molecular Weight (g/mol) 283.419
Chemical Name or Material AS 19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.