missing translation for 'onlineSavingsMsg'
Learn More

(aS)-a-Aminobenzenebutanoyl-L-leucyl-L-phenylalanine Methyl Ester Mono(trifluoroacetate), TRC

Product Code. 30317171
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317171 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317171 Supplier Toronto Research Chemicals Supplier No. TRCA5914251G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (AlphaS)-Alpha-Aminobenzenebutanoyl-L-leucyl-L-phenylalanine Methyl Ester Mono(trifluoroacetate)
CAS 868539-99-5
Molecular Formula C28H36F3N3O6
Quantity 1 g
Synonym (αS)-α-Aminobenzenebutanoyl-L-leucyl-L-phenylalanine Methyl Ester 2,2,2-Trifluoroacetate
InChI Formula InChI=1S/C26H35N3O4.C2HF3O2/c1-18(2)16-22(28-24(30)21(27)15-14-19-10-6-4-7-11-19)25(31)29-23(26(32)33-3)17-20-12-8-5-9-13-20;3-2(4,5)1(6)7/h4-13,18,21-23H,14-17,27H2,1-3H3,(H,28,30)(H,29,31);(H,6,7)/t21-,22-,23-;/m0./s1
SMILES COC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](N)CCc2ccccc2.OC(=O)C(F)(F)F
IUPAC Name methyl (2S)-2-[[(2S)-2-[[(2S)-2-amino-4-phenylbutanoyl]amino]-4-methylpentanoyl]amino]-3-phenylpropanoate;2,2,2-trifluoroacetic acid
Molecular Weight (g/mol) 567.6
Formula Weight 567.2556
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.