missing translation for 'onlineSavingsMsg'
Learn More

Ascarite™ II, CO2 absorbent, 20-30 mesh, capacity: 40-50%, product of Arthur H. Thomas Company

Product Code. 10481631 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10481631 500 g Glass Bottle 500g
10627504 100 g Glass Bottle 100g
15224684 2.5 kg Glass Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10481631 Supplier Thermo Scientific Acros Supplier No. 208095000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ascarite(II), CO2 absorbent, 20-30 mesh, capacity: 40-50%, CAS Number-81133-20-2, 1310-73-2, 7631-86-9, 100g, Beige to Brown, Danger, GHS H Statement: Causes severe skin burns and eye damage. | CAS: 81133-20-2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Gas Adsorbent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ascarite(II)™, CO2 absorbent, 20-30 mesh, capacity: 40-50%
Name Note 20- to 30-mesh
Color Beige to Brown
Physical Form Granular
CAS 81133-20-2, 1310-73-2, 7631-86-9
Quantity 500 g
MDL Number MFCD00132264
Packaging Glass Bottle
Particle Size Particle size analysis: 20 to 30 mesh
compliance-icons
Product Identifier
  • Ascarite(II), CO2 absorbent, product of Arthur H. Thomas Company
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.