missing translation for 'onlineSavingsMsg'
Learn More

Ascarite™ II, CO2 absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company

Product Code. 10092680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10092680 500 g 500g
10617504 100 g 100g
15214684 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10092680 Supplier Thermo Scientific Acros Supplier No. 208085000
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 81133-20-2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Gas Adsorbent
TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Ascarite(II)™, CO2 absorbent, 8 to 20 mesh
Name Note 8- to 20-mesh
Color Brown
Physical Form Granular
CAS 81133-20-2, 1310-73-2, 7631-86-9
Quantity 500 g
MDL Number MFCD00132264
Packaging Glass Bottle
Particle Size Particle size analysis: 8 to 20 mesh
compliance-icons
Identifiant produit
  • Ascarite(II)Ò, CO2 absorbent, product of Arthur H. Thomas Company
Mention d'avertissement
  • Danger
Catégorie de danger
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
Mention de danger
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
Informations supplémentaires
  • MIXTURE LIST-Contain: Sodium hydroxide

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.