missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Asiaticoside, 92+%

Product Code. 15822524 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15822524 5 g 5g
15812524 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15822524

Brand: Thermo Scientific Chemicals 461670050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16830-15-2 | C48H78O19 | 959.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16830-15-2
Molecular Formula C48H78O19
Molecular Weight (g/mol) 959.13
InChI Key WYQVAPGDARQUBT-NNUNPJGWSA-N
IUPAC Name 6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]oxan-2-yl]oxy}methyl)-3,4,5-trihydroxyoxan-2-yl (1S,2R,4aS,6aS,6bR,9R,10S,11R,12aR,14bS)-10,11-dihydroxy-9-(hydroxymethyl)-1,2,6a,6b,9,12a-hexamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylate
SMILES C[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CCC4[C@@]5(C)C[C@@H](O)[C@@H](O)[C@@](C)(CO)C5CC[C@@]34C)[C@@H]2[C@H]1C)C(=O)OC1OC(COC2OC(CO)C(OC3OC(C)C(O)C(O)C3O)C(O)C2O)C(O)C(O)C1O

Specifications

Melting Point 230.0°C to 233.0°C
Assay Percent Range 92.0 % min. (HPLC)
Quantity 5 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in warmed methanol
Formula Weight 959.12
Chemical Name or Material Asiaticoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.