missing translation for 'onlineSavingsMsg'
Learn More

Asiaticoside, MedChemExpress

Product Code. 16464837
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. Quantity unitSize
16464837 50 mg 50mg
16428988 10 mg 10mg
16409086 100 mg 100mg
3 options
This item is not returnable. View return policy

Product Code. 16464837

Brand: Medchem Express HYN0439

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Asiaticoside, a trisaccaride triterpene from Centella asiatica, suppresses TGF-β/Smad signaling through inducing Smad7 and inhibiting TGF-βRI and TGF-βRII in keloid fibroblasts; Asiaticoside shows antioxidant, anti-inflammatory, and anti-ulcer properties.

Asiaticoside, a trisaccaride triterpene from Centella asiatica, suppresses TGF-β/Smad signaling through inducing Smad7 and inhibiting TGF-βRI and TGF-βRII in keloid fibroblasts

Specifications

Chemical Name or Material Asiaticoside
CAS 16830-15-2
Color White
Purity Grade Notes Research
Molecular Formula C48H78O19
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (52.13 mM; Need ultrasonic)
SMILES C[C@@H]1CC[C@@]2(C(O[C@@H]3O[C@H](CO[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@]5([H])[C@H](O)[C@H](O)[C@@H](O)[C@H](C)O5)[C@@H](CO)O4)[C@@H](O)[C@H](O)[C@H]3O)=O)[C@@](C6=CC[C@@]([C@](C[C@@H](O)[C@H](O)[C@]7(CO)C)(C)[C@@]7([H])CC8)([H])[C@]8(C)[C@]6(C)CC2)([H])[C@H]1C
Molecular Weight (g/mol) 959.12
Formula Weight 959.12
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.