Learn More
Tocris Bioscience™ ASP 7663
GREENER_CHOICE
Selective TRPA1 activator
Brand:  Tocris Bioscience™ 5178/50
Encompass
Product Code. 15746546
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
ASP 7663 is a selective TRPA1 activator (EC50 values are ∽0.5 μM in human, mouse and rat). Displays little or no affinity at more than 60 other related receptors, channels and enzymes. Stimulates 5-HT release from QGP-1 cells. Exhibits an abdominal analgesic effect in vivo. Orally bioavailable.
Specifications
Specifications
ASP 7663
(2E)-2-[7-Fluoro-1,2-dihydro-1-(2-methylpropyl)-2-oxo-3H-indol-3-ylidene]acetic acid
Soluble to 100mM in DMSO and to 100mM in ethanol
Bioactive Small Molecule
1190217-35-6
50 mg
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research Use Only. Not For Human or Veterinary Use