missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Aspartame

Product Code. 16420898
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16420898 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16420898 Supplier MedChemExpress Supplier No. HYB0361
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Aspartame (SC-18862) is a methyl ester of a dipeptide. Aspartame can be used as a synthetic nonnutritive sweetener. Aspartame is composed of phenylalanine (50%), aspartic acid (40%) and methanol (10%).

Specifications

Purity 98.01%
CAS 22839-47-0
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 294.3 g/mol
Product Type Peptides
Quantity 5 g
Shipping Condition Room temperature
Solubility DMSO : 25 mg/mL (84.95 mM∣Need ultrasonic) ∣H2O : 5 mg/mL (16.99 mM∣Need ultrasonic)
Synonym SC-18862
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.