missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Aspirin

Product Code. 18035532
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18035532 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18035532 Supplier R&D Systems™ Supplier No. 4092/50
  • ONLINE EXCLUSIVE PRICE
    100.27 €
    90.30 € / 50mg
    Save 9.97 €
    9% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cyclooxygenase inhibitor; NSAID

Inhibitor of cyclooxygenase (COX)-1. Blocks the production of prostaglandins and thromboxanes. Exhibits antiplatelet and antithrombotic activities. Displays anticancer effects in some solid tumors. One of the first described NSAID drugs.
TRUSTED_SUSTAINABILITY

Specifications

CAS 50-78-2
Quantity 50 mg
Product Line Tocris
Molecular Formula C9H8O4
Formula Weight 180.16
Purity >99%
Molecular Weight (g/mol) 180.16
Chemical Name or Material Aspirin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.