missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals ASSURE H. pylori Rapid Test (CE)

Product Code. 31605227 Shop All MP Biomedicals Products
Change view
Click to view available options
Detectable Analytes:
H. pylori IgG
Unit Size:
20 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Unit Size
31605227 H. pylori IgG 20 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31605227 Supplier MP Biomedicals Supplier No. 43490020
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Immunochromatographic test device intended for the rapid detection of IgG antibodies to Helicobacter pylori in human serum, plasma or whole blood

  • The MP Diagnostics ASSURE H. pylori Rapid Test is an immunochromatographic test device intended for the rapid detection of IgG antibodies to Helicobacter pylori in human serum, plasma or whole blood.
  • It is intended as a clinical diagnostic test for diagnosing infection with H. pylori in patients with gastric disorders.
  • In addition, the presence of antibodies to a recombinant current infection marker (CIM) is indicative of current infection.
  • Usage Statement: For Sale Outside the US Only.

Specifications

CE Marker CE-IVD
Detectable Analytes H. pylori IgG
Type H. pylori Rapid Test
compliance-icons
Product Identifier
  • Rapid Test Kits
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.