missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals ASSURE HEV IgM Rapid Test (CE)

Product Code. 17362070 Shop All MP Biomedicals Products
Change view
Click to view available options
Detectable Analytes:
Hepatitis E Virus
Unit Size:
20 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Unit Size
17362070 Hepatitis E Virus 20 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17362070 Supplier MP Biomedicals Supplier No. 43160020
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

An IgM Rapid Test captures solid-phase immunochromatographic assay to detect hepatitis E virus.

  • All antibodies of IgM class, when present in the test sample, can be captured by anti-human IgM antibodies immobilized on the membrane.
  • The presence of HEV specific IgM antibodies can be differentially detected by a colloidal gold-labeled HEV antigen immobilized within the device, and can be visualized as pink/purple lines after assay.
  • In addition, immobilized rabbit IgG antibodies which can be recognized by colloidal gold-labeled anti-rabbit IgG antibodies were used as a control for proper function of the reagents.
  • Usage Statement: For Sale Outside the US Only.
TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes Hepatitis E Virus
Type IgM Rapid Test
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.