Learn More
Ansell AlphaTec™ 3000 Avant2 Airline Ultrasonically Welded and Taped - Model 756
Product Code. 17101085
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 4
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17101085 Large Pack of 4
17131075 Small Pack of 4
17141075 Small Pack of 4
17151075 Small Pack of 4
17161075 Medium Pack of 4
17171075 Medium Pack of 4
17191075 Large Pack of 4
17121085 X-Large Pack of 4
17131085 X-Large Pack of 4
17151085 2X-Large Pack of 4
17161085 2X-Large Pack of 4
17181085 3X-Large Pack of 4
17191085 3X-Large Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 17101085 Supplier Ansell Supplier No. YE30TA0075604
Estimated Shipment: 25-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
AlphaTec™ 3000 Avant2 Airline Ultrasonically Welded and Taped Suit features a panoramic visor design that ensures an unobstructed field of vision and a HEPA filter.
  • Specialized protection: The AlphaTec 3000 AVANT2 AIRline Ultrasonically Welded and Taped - Model 756 chemical protective suit has proven to protect wearers from over 200 chemicals.
  • Enhanced features: This full protective suit features a panoramic visor design that ensures an unobstructed field of vision
  • Genuine versatility: The AlphaTec 3000 AVANT2 AIRline Ultrasonically Welded and Taped - Model 756 full chemical protection suit also has an HEPA filter, which provides secondary protection from airline contamination and an SMC silencer keeps noise in the suit below 70 dB (even at maximum airflow)
  • Considered aesthetics: In addition, the attached socks with elasticated boot overflap was designed to be worn inside chemical protective boots with the leg overflap worn outside, to reduce the potential for chemical ingress
DESIGN FEATURES:
  • Fully encapsulated suit designed for use in combination with the permanently attached AlphaTec™ AVANT STS V.1 airline regulator.
  • Suit features include internal air-channel ventilation system double cuffs, finger and thumb loop, G01 AlphaTec™ 37-676 or G05 AlphaTec™ 29-500 gloves.
  • Attached boot ends with anti-slip PVC sole version also available and emergency rip cord.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Color Yellow
For Use With (Application) Opening and draining pumps valves or lines, Filling, blending and charging of raw materials, Contaminated Environments, Chemical Handling, Transferring liquids and solids, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes, Loading and unloading of proces equipment
Hood Type Fully Encapsulated
Material Multi-layer Barrier Laminate Fabric
Size Large
Ankle Style Elasticated Boot Overflap
Disposable Reusable
Gender Unisex
Product Type Chemical Protective Suit
Protection Class CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Seam Type Welded and Taped
Sterility Non-sterile