missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Astaxanthin

Product Code. 16488738
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488738 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488738 Supplier MedChemExpress Supplier No. HYB2163
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Astaxanthin, a red dietary carotenoid isolated from Haematococcus pluvialis, is a modulator of PPARγ and a potent antioxidant with antiproliferative, neuroprotective and anti-inflammatory activity. Astaxanthin has potential in the study of various diseases, such as cancers and Parkinson’s disease, cardiovascular disease. Due to its bright red colour, Astaxanthin could be used as a food colorant in animal feeds.

Specifications

Content And Storage -20°C, protect from light, stored under nitrogen∣The compound is unstable in solutions, freshly prepared is recommended.
Molecular Weight (g/mol) 596.84 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 472-61-7
Product Type Natural Products
Purity 98.0%
Solubility DMSO : 2 mg/mL (3.35 mM∣Need ultrasonic) ∣Acetone : <1 mg/mL (insoluble)
Target PPAR∣Reactive Oxygen Species∣STAT∣NF-κB∣Apoptosis
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.