missing translation for 'onlineSavingsMsg'
Learn More

Astaxanthin

Product Code. 10682862 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10682862 250 mg 250mg
10666374 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10682862 Supplier Thermo Scientific Acros Supplier No. 328612500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 472-61-7 | C40H52O4 | 596.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 472-61-7
Molecular Formula C40H52O4
Molecular Weight (g/mol) 596.85
MDL Number MFCD00672621
InChI Key MQZIGYBFDRPAKN-UWFIBFSHSA-N
Synonym astaxanthin, ovoester, astaxanthine, 3s,3's-astaxanthin, all-trans-astaxanthin, natupink, bioastin, carophyll pink, lucantin pink, trans-astaxanthin
PubChem CID 5281224
ChEBI CHEBI:40968
IUPAC Name (6S)-6-hydroxy-3-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-18-[(4S)-4-hydroxy-2,6,6-trimethyl-3-oxocyclohex-1-en-1-yl]-3,7,12,16-tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaen-1-yl]-2,4,4-trimethylcyclohex-2-en-1-one
SMILES C\C(\C=C\C=C(/C)\C=C\C1=C(C)C(=O)[C@@H](O)CC1(C)C)=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)C(=O)[C@@H](O)CC1(C)C

Specifications

Color Violet
Infrared Spectrum Authentic
Assay Percent Range 98+%
Quantity 250 mg
Formula Weight 596.86
Percent Purity ≥98.0%
Physical Form Powder
Chemical Name or Material Astaxanthin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.