missing translation for 'onlineSavingsMsg'
Learn More

Astragaloside I, MedChemExpress

Product Code. 16488978
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488978 10 mg 10mg
16431408 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488978 Supplier Medchem Express Supplier No. HYN0432
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Astragaloside I, one of the main active ingredients in Astragalus membranaceus, has osteogenic properties. Astragaloside I stimulates osteoblast differentiation through the Wnt/β-catenin signaling pathway.

Astragaloside I, one of the main active ingredients in Astragalus membranaceus, has osteogenic properties. Astragaloside I stimulates osteoblast differentiation through the Wnt/β-catenin signaling pathway.

Specifications

Chemical Name or Material Astragaloside I
CAS 84680-75-1
Color White
Purity Grade Notes Research
Molecular Formula C45H72O16
Quantity 10 mg
Synonym Astrasieversianin IV Cyclosieversioside B
Solubility Information DMSO : 50 mg/mL (57.53 mM; Need ultrasonic)
SMILES O[C@H]1[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@]([H])(O[C@@H]2C(C)(C)[C@@]([C@@H](O[C@@]3([H])[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O3)C[C@]4([H])[C@@]56CC[C@@]7(C)[C@@]4(C)C[C@H](O)[C@]7([H])[C@]8(C)O[C@H](C(C)(O)C)CC8)([H])[C@]5(C6)CC2)OC1
Molecular Weight (g/mol) 869.04
Formula Weight 869.04
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.