missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AT 1
GREENER_CHOICE

Product Code. 18738796
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18738796 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18738796 Supplier Tocris Bioscience™ Supplier No. 6356/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(+)-JQ1 based PROTAC with selectivity for BRD4

AT 1 is a cell penetrant Proteolysis Targeting Chimera (PROTAC®) compound based on (+)-JQ1 (Cat.No. 4499) conjugated to a von Hippel-Lindau (VHL) ligand. Rationally designed based on a ternary complex crystal structure to improve selectivity for BRD4 degradation compared to MZ1 (Cat.No. 6154). Demonstrates profound and selective degradation of BRD4 in cells at 1-3 μM, with negligible loss of BRD2 and BRD3.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S,4R)-1-(2R)-2-Acetamido-3-[[6-[2-[(6S)-4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl]acetamido]hexyl]thio]-3-methylbutanoyl]-4-hydroxy-N-[4-(4-methylthiazol-5-yl)benzyl)pyrrolidinine-2-carboxamide
CAS 2098836-45-2
Assay Percent Range >98%
Quantity 5 mg
Molecular Formula C48H58ClN9O5S3
Formula Weight Observed MW: 972.68
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.