Learn More
ATAGO™ Digital Urine Specific Gravity Refractometer, UG-α
Product Code. 11476278
Change view
Click to view available options
Urine Specific Gravity Refractometer
Product Code.
11476278 Urine Specific Gravity Refractometer
1 options
Product Code. 11476278 Supplier ATAGO™ Supplier No. 3464
Only null left
Add to basket
Edge
Add to basket
The ATAGO™ Digital Urine Specific Gravity Refractometer UG-α is an upgraded version of previous model UG-1. The minimum scale indication has been improved to display up to the fourth decimal.
  • Using the principles of light refraction, ATAGO Clinical Refractometers use special scales for clinical purposes such as specific gravity of urine
  • After pressing the START key, with the sample placed on the prism, the measurement values will be displayed on the LCD streaming as an electric message board
Specifications
Type Refractometer
Refractive Index 1.0000 to 1.0600
Model UG-α
Accuracy ± 0.0010
Resolution Min. Indication: 0.0001
Scale Type Urine Specific Gravity
Ambient Temperature (Metric) 10°C to 35°C