missing translation for 'onlineSavingsMsg'
Learn More

uvex Athletic All-Round Assembly Gloves

Product Code. 17250341 Shop All Uvex Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
10 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17250341 10 10 pairs
17210341 6 10 pairs
17220341 7 10 pairs
17230341 8 10 pairs
17240341 9 10 pairs
17737559 11 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17250341 Supplier uvex Supplier No. 6002810
  • ONLINE EXCLUSIVE PRICE
    41.95 €
    39.41 € / 10 pairs
    Save 2.54 €
    6% Off
View all promotions
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lightweight, dirt-resistant gloves are constructed from a blend of polyamide and elastane, with an NBR foam coating for enhanced grip capabilities.

  • Slim-fit provides tactile feedback when assembling smaller workpieces
  • NBR foam coating is abrasion resistant and enhances grip in dry or slightly damp environments
  • Dirt-resistant shell is ideal for mechanical tasks
  • Certified by OEKO-TEX™ Standard 100
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material NBR Foam
Color Gray, Black
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Elastane, Polyamide
Product Type Mechanical Protection Gloves
Size 10
EN388 Standard 4122X
For Use With (Application) Assembly, Packaging, Precision Work, and Transportation
Gender Unisex
Glove Cuff Style Knit
Series Athletic All-Round
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.