Learn More
uvex Athletic D5XP Safety Glove
Product Code. 17747559 Shop All Uvex Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
10 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17747559 6 10 pairs
17757559 7 10 pairs
17767559 8 10 pairs
17777559 9 10 pairs
17787559 10 10 pairs
17797559 11 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17747559 Supplier uvex Supplier No. 6003006
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
The Uvex athletic D5 XP cut protection gloves feature high level D cut protection and a reinforced thumb joint.
  • Outer material made from HPPE, steel and polyamide/elastane
  • Micro-NBR foam coating on palm and fingertips
  • High cut protection (Level D)
  • High mechanical resistance
  • Good grip on dry and (slightly) oily/wet workpieces
  • Flexible safety gloves with excellent fit
  • Good tactile feel
  • Safety glove with knitted cuff and reinforced thumb joint
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 388 (4 X 4 3 D)
Coating Material Nitrile Foam
Color Gray, Black
Hand Ambidextrous
Latex Free Yes
Material HPPE, Steel Fiber, Nylon, Elastane
Product Type Cut-resistant Gloves
Size 6