missing translation for 'onlineSavingsMsg'
Learn More

uvex Athletic Lite Assembly Gloves

Product Code. 17290331 Shop All Uvex Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
10 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17290331 10 10 pairs
17727559 6 10 pairs
17260331 7 10 pairs
17270331 8 10 pairs
17280331 9 10 pairs
17200341 11 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17290331 Supplier uvex Supplier No. 6002710
  • ONLINE EXCLUSIVE PRICE
    41.95 €
    39.41 € / 10 pairs
    Save 2.54 €
    6% Off
View all promotions
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are constructed from a combination of polyamide and elastane, with a nitrile rubber microfoam coating for enhanced grip in dry or slightly damp applications.

  • Slim, ergonomic fit allows for precision work with high levels of user comfort
  • Fine-structured, open-pored microfoam coating enhances grip capabilities
  • Breathable, matte coating reduces excessive perspiration
  • Ideal for precision assembly work, maintenance, inspection, and sorting
  • Certified by OEKO-TEX™ Standard 100
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material NBR MicroFoam
Color Blue, Black
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Polyamide, Elastane
Product Type Mechanical Protection Gloves
Size 10
EN388 Standard 4132X
For Use With (Application) Inspection, Precision Assembly Work, Repair Work, and Sorting
Gender Unisex
Glove Cuff Style Knit
Series Athletic Lite
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.