missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Atipamezole hydrochloride

Product Code. 18074822
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18074822 10 mg 10mg
18024292 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18074822 Supplier R&D Systems™ Supplier No. 2937/10
  • ONLINE EXCLUSIVE PRICE
    332.51 €
    288.75 € / 10mg
    Save 43.76 €
    13% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective α2 antagonist

Selective α2-adrenergic receptor antagonist (Ki values are 1.1, 1.0, 0.89, 1300 and 6500 nM for α2A, α2B, α2C, α1A and α1B receptors respectively). Brain penetrant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104075-48-1
Molecular Formula C14H17ClN2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 248.75
Purity >99%
Molecular Weight (g/mol) 248.75
Chemical Name or Material Atipamezole hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.