missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ATN 161

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
ATN 161 is a α5β1 integrin receptor antagonist. Reduces liver metastasis and improves survival in combination with 5-FU (Cat. No. 3257) in a mouse model of colon cancer. Reduces MLL tumor growth in rats and decreases tumor volume of human MDA-MB-231 cell xenografts in mice.
Specifications
Specifications
| CAS | 262438-43-7 |
| Content And Storage | Store at −20°C |
| For Use With (Application) | α5β1 integrin receptor antagonist |
| Lyophilized | Yes |
| Molecular Weight (g/mol) | 597.65 |
| Product Type | Peptide |
| Quantity | 10 mg |
| Sequence | PHSCN |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction