missing translation for 'onlineSavingsMsg'
Learn More

Melford ATP

Product Code. 30018026
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018026 1 g 1g
30018028 5 g 5g
30018027 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018026 Supplier Melford Supplier No. A300301.0
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An essential nucleotide triphosphate with a variety of roles in energy metabolism, signal transduction and nucleic acid metabolism.

An essential nucleotide triphosphate with a variety of roles in energy metabolism, signal transduction and nucleic acid metabolism.

Specifications

Quantity 1 g
Molecular Formula C10H14N5Na2O13P3-3H2O
CAS 51963-61-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.