missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford ATP
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
Description
An essential nucleotide triphosphate with a variety of roles in energy metabolism, signal transduction and nucleic acid metabolism.
An essential nucleotide triphosphate with a variety of roles in energy metabolism, signal transduction and nucleic acid metabolism.
Specifications
Specifications
| Quantity | 10 g |
| Molecular Formula | C10H14N5Na2O13P3-3H2O |
| CAS | 51963-61-2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction