missing translation for 'onlineSavingsMsg'
Learn More

Melford ATP

Product Code. 30018028
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 30018028

Brand: Melford A300305.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An essential nucleotide triphosphate with a variety of roles in energy metabolism, signal transduction and nucleic acid metabolism.

An essential nucleotide triphosphate with a variety of roles in energy metabolism, signal transduction and nucleic acid metabolism.

Specifications

Quantity 5 g
Molecular Formula C10H14N5Na2O13P3-3H2O
CAS 51963-61-2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.