missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress ATP-polyamine-biotin

Product Code. 16492409
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
100 mg
Unit Size:
1mg
2mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492409 1 mg 1mg
16490376 100 mg 2mg
16421168 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16492409 Supplier MedChemExpress Supplier No. HYD0183
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ATP-polyamine-biotin, the first cell-permeable ATP analogue, is an efficient kinase cosubstrate. ATP-polyamine-biotin promotes biotin labeling of kinase substrates in live cells.

Specifications

Quantity 1 mg
Product Type Biochemical Assay Reagents
Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
CAS 1800401-93-7
Molecular Weight (g/mol) 945.85 g/mol
Solubility H2O : 6 mg/mL (6.34 mM; Need ultrasonic and warming)
Target HBV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.