missing translation for 'onlineSavingsMsg'
Learn More

Atracurium besylate, MedChemExpress

Product Code. 16498216
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498216 100 mg 100mg
16492086 500 mg 500mg
16434327 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498216 Supplier Medchem Express Supplier No. HYB0292A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Atracurium (BW-33A) besylate is a potent, competitive and non-depolarizing neuromuscular blocking agent. Atracurium besylate also is an AChR receptor antagonist. Atracurium besylate induces bronchoconstriction and neuromuscular blockade. Atracurium besylate promotes astroglial differentiation.

Atracurium (BW-33A) besylate is a potent, competitive and non-depolarizing neuromuscular blocking agent. Atracurium besylate also is an AChR receptor antagonist. Atracurium besylate induces bronchoconstriction and neuromuscular blockade. Atracurium besylate promotes astroglial differentiation.

Specifications

Chemical Name or Material Atracurium besylate
CAS 64228-81-5
Color White
Purity Grade Notes Research
Molecular Formula C65H82N2O18S2
Quantity 100 mg
Synonym BW-33A
Solubility Information DMSO : 100 mg/mL (80.42 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (26.80 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC=C1)([O-])=O.O=C(OCCCCCOC(CC[N+]2(C)C(CC3=CC=C(OC)C(OC)=C3)C4=C(C=C(OC)C(OC)=C4)CC2)=O)CC[N+]5(C)C(CC6=CC=C(OC)C(OC)=C6)C7=C(C=C(OC)C(OC)=C7)CC5.O=S(C8=CC=CC=C8)([O-])=O
Molecular Weight (g/mol) 1243.48
Formula Weight 1243.48
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.