missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Attune™ Autosampler Syringe

Product Code. 15787597 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15787597 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15787597 Supplier Applied Biosystems™ Supplier No. 4478686
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This is a replacement syringe for the Attune™ Auto Sampler.

  • Replacement syringe for the Attune™ Auto Sampler
TRUSTED_SUSTAINABILITY

Specifications

Product Line Attune
Content And Storage Store at room temperature
Quantity 1 mL
Shipping Condition Room Temperature
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Sampler Syringe

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.