missing translation for 'onlineSavingsMsg'
Learn More

Attune™ CytPix™ Violet 6 Conversion Kit, for red laser

Product Code. 17391929
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17391929

Brand: Invitrogen A50928

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to reconfigure Attune NxT four-channel violet instruments to allow detection of six channels of violet. A conversion kit is required for each original, and for any upgraded lasers of the instrument.

  • Designed to reconfigure Attune NxT four-channel violet instruments to allow detection of six channels of violet
  • A conversion kit is required for each original, and for any upgraded lasers of the instrument
  • Conversion is carried out by a field service engineer
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Flow Cytometry
For Use With (Equipment) Attune CytPix Flow Cytometer
Includes Filters, dichroics
Product Line DJM
Quantity 1 Kit
Regulatory Status RUO
Shipping Condition RT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.