missing translation for 'onlineSavingsMsg'
Learn More

Attune™ Focusing Fluid (1X)

Product Code. 10019478
Click to view available options
Quantity:
1 L
10 L
20 L
6 x 1 L
Unit Size:
1 L
10 L
20 L
Pack of 6
This item is not returnable. View return policy

Product Code. 10019478

Brand: Invitrogen 4449791

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Attune™ Focusing Fluid (1X)

Attune™ Focusing Fluid is an optimized buffer solution that is used in the Attune™ Acoustic Focusing Cytometer for focusing and transportation of particles throughout the fluidics system.

Maintains System Cleanliness
Attune™ Focusing Fluid is a sterile, buffered, azide-free support reagent that contains a surfactant that helps clean the fluidic lines of the Attune™ Acoustic Focusing Cytometer. The Attune™ Focusing Fluid was designed to minimize bubble formation and to prevent build-up of cellular material in the flow cell, resulting in less instrument clogging.

Convenient
The ready-to-use 1X focusing fluid was designed to eliminate the need for dilution prior to use, as well as access to a purified water system. The convenient 10 L packaging provides enough focusing fluid to operate for at least 10 days (the average focusing fluid consumption per day is less than 1 L) and has a spigot to to make filling of the onboard fluidics tanks easier.

High Quality
Attune™ Focusing Fluid is prepared, filtered, and packaged in a sterile environment to ensure the highest quality. Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.
TRUSTED_SUSTAINABILITY

Spezifikation

Content And Storage Store at 15—30°C
For Use With (Equipment) Attune™ Acoustic Focusing Cytometer
Format Liquid
Product Line Attune
Quantity 6 x 1 L
Product Type Focusing Fluid

For Research Use Only. Not for use in diagnostic procedures.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt