missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT Automation Bundle, hotels

Product Code. 15965041
Change view
Click to view available options
Quantity:
1 bundle
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15965041 1 bundle 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15965041 Supplier Invitrogen™ Supplier No. A33008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Attune NxT Automation Bundle consists of an Orbitor RS2 Microplate Mover and a barcode reader for quick and easy plate identification.

This Orbitor RS2 Microplate Mover is an automation robot designed for use with the Attune NxT Acoustic Focusing Cytometer that enables flow cytometry in automated environments for increased sample throughput. It is offered in three different configurations: one with sequential access stacks for plate storage and handling (Cat. No. A33007), one with random access hotels (24 plates/3 hotels) (this page), and one with two sequential access stacks and one random access hotel (Cat. No. A35220).

Extensive vertical reach allows the Orbitor RS2 Microplate Mover to access virtually any microplate instrument location, including stacked instruments, and its telescoping arm can reach a variety of distances. The ingenious arm travels through the base, resulting for rapid and efficient movements. This robot has a full 360°workspace with unlimited base rotation; this eliminates the need for hard stops and reverse movements.

Advanced closed-loop motion control provides collision detection and position monitoring, allowing the Orbitor RS2 Microplate Mover to quickly resume without the need to re-home. With a wide range of grip-control functionality, this robot combines performance with innovation. From variable force control to plate detection, the feature set is designed to impress. The result is remarkable speed, precise motion, and consistent, accurate plate placement.

The Orbitor RS2 Microplate Mover is powered by Thermo Scientific Momentum Integration Software with features such as Momentum scheduling software, error detection and recovery, and a dynamic scheduler that allows changes while running.

TRUSTED_SUSTAINABILITY

Specifications

Product Line Attune
Content And Storage • Orbitor RS2 Microplate Mover
• Integrated barcode reader
• 3 Plates hotels
• Momentuum software

Quantity 1 bundle
Shipping Condition Room Temperature
For Use With (Equipment) Attune NxT Acoustic Focusing Cytometer
Product Type Automation Bundle

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.