missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT and CytPix Drip Tray

Product Code. 16576250
Change view
Click to view available options
Quantity:
1 tray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16576250 1 tray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16576250 Supplier Invitrogen™ Supplier No. 100023101
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Attune NxT Drip Tray is an optional accessory for the Attune NxT Acoustic Focusing Cytometer that helps prevent sample from spilling onto the laboratory bench if the sample tube becomes dislodged from the SIP tube.

  • The drip tray has an indented lip that will hold several milliliters of spilled sample
TRUSTED_SUSTAINABILITY

Specifications

Product Line Attune
Content And Storage Store at room temperature.

Quantity 1 tray
Shipping Condition Room Temperature
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Drip Tray

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.