missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT Level Sensor Cable

Product Code. 17146088
Click to view available options
Quantity:
1 cable
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17146088

Brand: Invitrogen™ 100087973CST

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This is a replacement level sensor cable for the focusing fluid and waste bottles of the Attune NxT Acoustic Focusing Cytometer

  • This is a replacement level sensor cable for the focusing fluid and waste bottles of the Attune NxT Acoustic Focusing Cytometer.
  • It can be replaced easily without replacing the entire bottle assembly if the sensor connector is broken.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 Cable
For Use With (Equipment) Attune NxT
Format Sensor cable
Product Line FPL
Quantity 1 cable
Shipping Condition Room Temperature
Product Type Flow Cytometer Accessory
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.