missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT Software 21 CFR Part 11

Product Code. 17194749
Change view
Click to view available options
Quantity:
1 License
1 Server license, 10 users
1 Server license, 5 users
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17194749 1 License Each
17138238 1 Server license, 5 users Each
17148238 1 Server license, 10 users Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17194749 Supplier Invitrogen™ Supplier No. A47288
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Attune NxT Software 21 CFR Part 11 is for use with the Attune NxT Acoustic Focusing Cytometer and supports compliance with 21 CFR Part 11 FDA guidelines for security, auditing, and electronic (SAE) signatures using an SAE console.

  • The software helps prevent unauthorized access to the system and is available for use on computers with the Windows 10 operating system.
  • Authentication of user log-in information
  • Ability to set password guidelines and expiry dates
  • Lockout access and re-authentication requirements after a period of inactivity
  • System lockout upon unauthorized attempts to access software
  • Ability of administrator to define user roles and permissions
  • Identification of data tampering
  • Generation of audit trail log
  • Ability of administrator to determine functions required to input reason for change
  • Tracking of changes to user roles, account management, and system settings
  • Assignment of unique identifiers for experiments and objects within experiments to identify any changes from original record
  • Audit trail record presented in human-readable format
  • Administrator determination of e-signature requirements
  • Ability to review records and sign electronically
  • Along with the 21 CFR part 11 software license, our service and support organization offers these services to help you validate you instrument, software, and computer: IQOQ and computer software validation services.
TRUSTED_SUSTAINABILITY

Spécification

For Use With (Equipment) Attune™ NxT Acoustic Focusing Cytometer
Product Line Attune
Product Type Flow Cytometer Software
Quantity 1 License
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.