missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT Violet 6 Conversion Kit

Product Code. 31941518
Change view
Click to view available options
Color:
Blue
Red
Violet
Yellow
Unit Size:
1 set
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31941518 Blue Each
31722852 Violet 1 set
31722922 Red 1 set
31722771 Yellow 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31941518 Supplier Invitrogen™ Supplier No. A35428
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Attune NxT Violet 6 Conversion kits are designed to reconfigure Attune NxT four-channel violet instruments to allow detection of six channels of violet.

Attune NxT Violet 6 Conversion kits are for use in the reconfiguration of four-channel Attune NxT instruments to enable detection of six channels of violet. A conversion kit is required for each original and any upgraded lasers of the instrument. The conversion is carried out in your laboratory by a Field Service Engineer.

TRUSTED_SUSTAINABILITY

Specifications

Product Line Attune
Content And Storage Optical block and filters
Quantity 1 Kit
Color Blue
Shipping Condition Room Temperature
For Use With (Equipment) Attune NxT Four-Channel Violet Instrument
Product Type Conversion Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.